missing translation for 'onlineSavingsMsg'
Learn More

Dibutylgermanium dichloride, 98%, Thermo Scientific™

£212.00

Chemical Identifiers

CAS 4593-81-1
Molecular Formula C8H18Cl2Ge
Molecular Weight (g/mol) 257.73
MDL Number MFCD00013590
InChI Key VKMPTHGEGBKPBQ-UHFFFAOYSA-N
Synonym di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
PubChem CID 20723
IUPAC Name dibutyl(dichloro)germane
SMILES CCCC[Ge](CCCC)(Cl)Cl
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10433073
SDS View Documents
Thermo Scientific Acros
381110050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

4593-81-1
257.73
VKMPTHGEGBKPBQ-UHFFFAOYSA-N
20723
CCCC[Ge](CCCC)(Cl)Cl
C8H18Cl2Ge
MFCD00013590
di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
dibutyl(dichloro)germane

Specifications

4593-81-1
1.2080g/mL
Authentic
C8H18Cl2Ge
1.4720 to 1.474
1.208
VKMPTHGEGBKPBQ-UHFFFAOYSA-N
dibutyl(dichloro)germane
20723
98%
Dibutylgermanium dichloride
Colorless
242°C
97.5% min. (GC)
(CH3CH2CH2CH2)2Ge(Cl)2
MFCD00013590
di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
CCCC[Ge](CCCC)(Cl)Cl
257.73
257.73
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 224-986-9

Product Identifier
  • Dibutylgermanium dichloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.