missing translation for 'onlineSavingsMsg'
Learn More

Diallyldibutyltin, Thermo Scientific™

£18.28 - £168.00

Chemical Identifiers

CAS 15336-98-8
Molecular Formula C14H28Sn
Molecular Weight (g/mol) 315.088
MDL Number MFCD00014944
InChI Key LCGVYMRFNOWPGQ-UHFFFAOYSA-N
Synonym diallyldibutyltin, diallyldibutylstannane, dibutylbis prop-2-en-1-yl stannane, diallyl dibutyl stannane, diallyldi-n-butyltin, acmc-1bvvj, dibutyl-bis prop-2-enyl stannane
PubChem CID 84873
IUPAC Name dibutyl-bis(prop-2-enyl)stannane
SMILES CCCC[Sn](CCCC)(CC=C)CC=C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354567
View Documents
Thermo Scientific Alfa Aesar
039621.02
0.5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11334567
View Documents
Thermo Scientific Alfa Aesar
039621.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11344567
View Documents
Thermo Scientific Alfa Aesar
039621.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

15336-98-8
315.088
LCGVYMRFNOWPGQ-UHFFFAOYSA-N
84873
CCCC[Sn](CCCC)(CC=C)CC=C
C14H28Sn
MFCD00014944
diallyldibutyltin, diallyldibutylstannane, dibutylbis prop-2-en-1-yl stannane, diallyl dibutyl stannane, diallyldi-n-butyltin, acmc-1bvvj, dibutyl-bis prop-2-enyl stannane
dibutyl-bis(prop-2-enyl)stannane

Specifications

15336-98-8
81°C (0.2 mmHg)
1.5
UN2788
diallyldibutyltin, diallyldibutylstannane, dibutylbis prop-2-en-1-yl stannane, diallyl dibutyl stannane, diallyldi-n-butyltin, acmc-1bvvj, dibutyl-bis prop-2-enyl stannane
LCGVYMRFNOWPGQ-UHFFFAOYSA-N
dibutyl-bis(prop-2-enyl)stannane
84873
Liquid
1.1002 g/mL
C14H28Sn
MFCD00014944
Air and moisture sensitive
Insoluble in water.
CCCC[Sn](CCCC)(CC=C)CC=C
315.088
315.07
Diallyldibutyltin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 239-368-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications