missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyltin dichloride, 98%, Thermo Scientific Chemicals

£38.99 - £104.00

Chemical Identifiers

CAS 19429-30-2
Molecular Formula C8H18Cl2Sn
Molecular Weight (g/mol) 303.842
MDL Number MFCD00009854
InChI Key PEGCFRJASNUIPX-UHFFFAOYSA-L
Synonym di-tert-butyldichlorostannane, di-tert-butyltin dichloride, di-t-butyltin dichloride, ditert-butyl dichloro stannane, acmc-1bpuo, di-tert-butylzinndichlorid
PubChem CID 88053
IUPAC Name ditert-butyl(dichloro)stannane
SMILES CC(C)(C)[Sn](C(C)(C)C)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324567
View Documents
Thermo Scientific Alfa Aesar
039619.02
0.5 g
£38.99
0.50g
Please to purchase this item. Need a web account? Register with us today!
11314567
View Documents
Thermo Scientific Alfa Aesar
039619.04
2 g
£104.00
2g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19429-30-2
303.842
PEGCFRJASNUIPX-UHFFFAOYSA-L
88053
CC(C)(C)[Sn](C(C)(C)C)(Cl)Cl
C8H18Cl2Sn
MFCD00009854
di-tert-butyldichlorostannane, di-tert-butyltin dichloride, di-t-butyltin dichloride, ditert-butyl dichloro stannane, acmc-1bpuo, di-tert-butylzinndichlorid
ditert-butyl(dichloro)stannane

Specifications

19429-30-2
White
>110°C (230°F)
C8H18Cl2Sn
MFCD00009854
Moisture sensitive
PEGCFRJASNUIPX-UHFFFAOYSA-L
ditert-butyl(dichloro)stannane
88053
98%
Di-tert-butyltin dichloride
42°C to 43°C
66°C (3 mmHg)
Acrid
[(CH3)3C]2SnCl2
UN2928
di-tert-butyldichlorostannane, di-tert-butyltin dichloride, di-t-butyltin dichloride, ditert-butyl dichloro stannane, acmc-1bpuo, di-tert-butylzinndichlorid
CC(C)(C)[Sn](C(C)(C)C)(Cl)Cl
303.842
303.83
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311+H331-H314

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 243-051-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only