missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Di-n-butylammonium tetrafluoroborate, 98%

Product Code. 11499726
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 12107-76-5
Molecular Formula C8H20BF4N
Molecular Weight (g/mol) 217.06
MDL Number MFCD00034899
InChI Key CAZKFGXFAVXKIF-UHFFFAOYSA-O
Synonym di-n-butylammonium tetrafluoroborate, dibutylazanium tetrafluoroborate, dibutylamine tetrafluoroborate, dibutylammonium tetrafluoroborate 1-, dibutylammonium tetrafluoroborate, dibutylammonium, tetrafluoroborate, dibutylamine, tetrafluoroborate 1-, 1-butanamine, n-butyl-, tetrafluoroborate 1-, dibutylamine tetrafluoborate, c8h20n.bf4
PubChem CID 25510
IUPAC Name dibutylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCC[NH2+]CCCC

Specifications

Linear Formula (C4H9)2NH2BF4
Quantity 100 g
Formula Weight 217.06
Percent Purity 98%
Chemical Name or Material Di-n-butylammonium tetrafluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.