missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Raffinose pentahydrate, 99+%

Catalog Number p-3737320
73.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
17629-30-0
C18H32O16·5H2O
594.52
MFCD00071590
BITMAWRCWSHCRW-ORBWWOJFSA-N
d-+-raffinosepentahydrate
134129414
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O
17629-30-0
C18H32O16·5H2O
594.52
MFCD00071590
BITMAWRCWSHCRW-ORBWWOJFSA-N
d-+-raffinosepentahydrate
134129414
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 78.0°C to 80.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 17, V,8, 403
Merck Index 14, 8096
Specific Rotation Condition + 105.00 (20.00°C c=10,water,dry basis)
Specific Rotation + 105.00
Solubility Information (10% in water) Clear colorless
Formula Weight 594.52
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Raffinose pentahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'