missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals α-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%

Product Code. 10506081
Click to view available options
Quantity:
50 mg
500 mg
2.5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19046-60-7
Molecular Formula C6H11K2O9P
Molecular Weight (g/mol) 336.32
MDL Number MFCD00065475
InChI Key KCIDZIIHRGYJAE-YGFYJFDDSA-L
Synonym alpha-d-galactose-1-phosphate dipotassium salt dihydrate
PubChem CID 131875454
IUPAC Name potassium;[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
SMILES [K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Color Colorless to White
CAS Min % 97.5
CAS Max % 100.0
Physical Form Crystalline Powder and/or Crystals
Quantity 500 mg
Assay Percent Range 98%
Solubility Information (5% in water) Clear colorless
Formula Weight 426.39
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Appearance (5% aq. soln.) clear, colorless
Chemical Name or Material α-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%
Show More Show Less
Product Identifier
  • a-D-Galactose 1-phosphate dipotassium salt pentahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.