missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals

£26.03 - £121.61

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15229315
View Documents
Thermo Scientific Acros
451962500
250 g
£26.03
250g
Please to purchase this item. Need a web account? Register with us today!
15209315
View Documents Promotion Details
Thermo Scientific Acros
451960010
1 kg
£67.03
1kg
Please to purchase this item. Need a web account? Register with us today!
15219315
View Documents
Thermo Scientific Acros
451960050
5 kg
£121.61
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57-48-7
180.16
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O
C6H12O6
BJHIKXHVCXFQLS-UYFOZJQFSA-N
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one

Specifications

57-48-7
0.5% max
15,4302
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
180.16
103.0°C to 105.0°C
C6H12O6
− 92.25
Solubility in water: 3750g/L (20°C). Other solubilities: soluble in alcohol,pyridine,ether,ethylamine,and methylamine
C(C(C(C(C(=O)CO)O)O)O)O
180.16
CHEBI:48095
D(-)-Fructose
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-333-3

Product Identifier
  • D(-)-Fructose

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.