Learn More
Thermo Scientific Chemicals D-(+)-Cellobiose, 98+%
Chemical Identifiers
CAS | 528-50-7 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.30 |
MDL Number | MFCD00136034 |
InChI Key | GUBGYTABKSRVRQ-UHFFFAOYNA-N |
Synonym | d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose |
PubChem CID | 57370084 |
SMILES | OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11431938
|
Thermo Scientific Chemicals
A14553.06 |
5 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441938
|
Thermo Scientific Chemicals
A14553.14 |
25 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451938
|
Thermo Scientific Chemicals
A14553.22 |
100 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.
Solubility
Soluble in water (1 g/8ml). Insoluble in ether and ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
528-50-7 | |
342.30 | |
GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
57370084 |
C12H22O11 | |
MFCD00136034 | |
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Specifications
528-50-7 | |
≥98% | |
MFCD00136034 | |
14,1961 | |
Soluble in water (1g/8ml). Insoluble in ether and ethanol. | |
+34° (c=10 in Water, 15h) | |
(3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
57370084 | |
≥98% |
234°C (decomposition) | |
C12H22O11 | |
93795 | |
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
342.30 | |
342.3 | |
D-(+)-Cellobiose |
Safety and Handling
missing translation for 'einecsNumber' : 208-436-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.