missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cystatin C Rat ELISA Kit

Sandwich ELISA Kit

£530.00 - £2310.00

Specifications

Accession Number P14841
Assay Sensitivity 1.2 ng/mL
Calibration Range 1.2 ng/mL to 750 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate Biotin
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405373
View Documents
Invitrogen™
ERCST3
96 Tests
£530.00
1 set
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
17175909
View Documents
Invitrogen™
ERCST3X5
5 x 96 Tests
£2310.00
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
Description

Description

The Rat Cystatin C (CST3) ELISA quantitates Rt Cystatin C in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt Cystatin C. Principle of the method The Rat Cystatin C solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Cystatin C is a cysteine (thiol) protease inhibitor that belongs to the type II cystatin gene superfamily and is the most abundant extracellular inhibitor of cysteine proteases. Cystatin C is a constitutively secreted, amyloidogenic protein, which forms a two-fold symmetric dimer and modulates both cysteine protease activity and the expression of class II MHC molecules. Expression of cystatin C is an indicator of kidney function and glomerular filtration rate. Mutations in the cystatin C gene can lead to protein aggregates, which are implicated in hereditary amyloid angiopathy (HCCAA) and cerebral hemorrhage. Although both wild-type and mutant cystatin C are capable of forming concentration dependent inactive dimers, mutant cystatin C dimerizes at lower concentrations and is more susceptible to serine proteases, which may facilitate aggregation. In neuronal cells, oxidative stress stimulates expression of cystatin C, which may positively regulate apoptosis.
Specifications

Specifications

P14841
1.2 ng/mL to 750 ng/mL
Biotin
Proteases, Peptidases & Inhibitors
This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 , beta-NGF, TIMP-1, TNF-alpha .
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
25307
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma, 25 μL; Serum, 25 μL; Supernatant, 100 μL
CYSC
1 hr. 20 min.
1.2 ng/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
Colorimetric Microplate Reader
Cst3
<10%
HRP
Apoptosis
2°C to 8°C
Rat
4 hr. 45 min.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.