missing translation for 'onlineSavingsMsg'
Learn More

Cylinder Brush

Manufactured with galvanized steel handle. Pinnacle Brush™ Cylinder Brush features nylon bristles.

Brand:  PINNACLE Brush T2403

Code : 03

Additional Details : Weight : 1.26000kg

Product Code. 12303745

  • £165.22 / Pack of 10
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

  • Fan-shaped end
  • Galvanised steel wire handle
Specifications

Specifications

Cylinder Brush
Steel Wire
685 mm
10 Pk.
Nylon
64 mm
150 mm
64 x 150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers