Learn More
Cyclohexanone, ACS, 99+%, Thermo Scientific Chemicals
£27.44 - £168.84
Chemical Identifiers
CAS | 108-94-1 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.145 |
MDL Number | MFCD00001625 |
InChI Key | JHIVVAPYMSGYDF-UHFFFAOYSA-N |
Synonym | ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon |
PubChem CID | 7967 |
ChEBI | CHEBI:17854 |
IUPAC Name | cyclohexanone |
SMILES | C1CCC(=O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474990
|
Thermo Scientific Alfa Aesar
033309.AP |
500 mL |
£27.44
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387628
|
Thermo Scientific Alfa Aesar
033309.K2 |
1 L |
£48.94
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307638
|
Thermo Scientific Alfa Aesar
033309.M1 |
4 x 1 L |
£168.84
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397628
|
Thermo Scientific Alfa Aesar
033309.K7 |
4 L |
£133.67
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-94-1 | |
98.145 | |
JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
7967 | |
cyclohexanone |
C6H10O | |
MFCD00001625 | |
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
CHEBI:17854 | |
C1CCC(=O)CC1 |
Specifications
°C to 26°C | |
Colorless | |
0.947 g/cm3 at 20°C | |
47°C (116°F) | |
1.4507 | |
C6H10O | |
4.5 hPa (3mm Hg) at 20°C | |
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
cyclohexanone | |
7967 | |
98.15 | |
Cyclohexanone, For ACS analysis |
108-94-1 | |
Colorless liquid | |
154°C to 156°C | |
Liquid | |
>99% | |
MFCD00001625 | |
385735 | |
Miscible with water and ethanol,acetone,ethyl ether and other organic solvents. | |
C1CCC(=O)CC1 | |
98.145 | |
CHEBI:17854 | |
ACS |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H318-H335-H336
missing translation for 'dotInformation' : DOT Class: 3; Packing Group: III
missing translation for 'einecsNumber' : 203-631-1
missing translation for 'rtecsNumber' : GW1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.