Learn More
Chemical Identifiers
CAS | 110-82-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00003814 |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
PubChem CID | 8078 |
ChEBI | CHEBI:29005 |
IUPAC Name | cyclohexane |
SMILES | C1CCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10539633
|
Thermo Scientific Acros
210570010 |
1 L | Glass Bottle |
£27.84
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11316185
|
Thermo Scientific Acros
210570025 |
2.5 L | Glass Bottle |
£50.45
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924431
|
Thermo Scientific Acros
210570100 |
10 L | Plastic Drum |
£147.74
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Specifications
6.5°C | |
0.7700g/mL | |
−18°C | |
Liquid | |
99.5% | |
0.77 | |
C6H12 | |
0.94 mPa.s (20°C) | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
84.15 | |
Extra Pure |
110-82-7 | |
81.0°C | |
Authentic | |
1.4250 to 1.4270 | |
0.005% max. | |
C6H12 | |
MFCD00003814 | |
0.01% max. | |
Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4. | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 | |
99.5% | |
Cyclohexane |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Danger
- Cyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only