missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR4 Mouse anti-Human, Clone: 4B5E4, Proteintech
Mouse Monoclonal Antibody
£132.00 - £343.00
Specifications
Antigen | CXCR4 |
---|---|
Clone | 4B5E4 |
Concentration | 1.5 mg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816995
|
Proteintech
60042-1-IG-20UL |
20 μL |
£132.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806995
|
Proteintech
60042-1-IG-150UL |
150 μL |
£343.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR4 may be involved in WHIM syndrome (WHIMS). Immunodeficiency disease characterized by neutropenia, hypogammaglobulinemia and extensive human papillomavirus (HPV) infection. Despite the peripheral neutropenia, bone marrow aspirates from affected individuals contain abundant mature myeloid cells, a condition termed myelokathexis. The disease is caused by mutations affecting the gene represented in this entry.Specifications
CXCR4 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD184, CXC R4, CXCR 4, CXCR4, CXCR4B, D2S201E, FB22, Fusin, HM89, HSY3RR, LAP3, LCR1, LESTR, SDF 1 receptor, WHIM | |
CXCR4 | |
IgM | |
Antigen Affinity Chromatography | |
Antibody |
4B5E4 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
P61073 | |
7852 | |
CXCR4 Fusion Protein Ag1528 | |
Primary | |
-20°C | |
CXCR4 |