Learn More
Thermo Scientific™ Culti-Loops™ Streptococcus agalactiae group B CL 810
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4607005
Product Code. 12965733
  • £182.00 / Pack of 5
Explore more special offers
Description
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Streptococcus agalactiae group B CL 810
Culti-Loops Quality Control Organism
Qualitative QC
Culti-Loops
Cocci
5 Loops/Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.