Learn More
Thermo Scientific™ Culti-Loops™ Haemophilus influenzae ATCC™ 35056™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4603840
201.19 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
35056TM* | |
Culti-Loops | |
Fastidious | |
5 Loops/Pack |
Haemophilus influenzae ATCC™ 35056™; Beta-lactamase positive | |
Culti-Loop | |
Qualitative QC | |
WDCM00037, [CIP 103777] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.