Learn More
Thermo Scientific™ Culti-Loops™ Naganishia albida var. albidus ATCC™ 10666™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4601811
174.32 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12958928
  • £182.00 / Pack of 5
Explore more special offers
Description
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10666TM*
Culti-Loops Quality Control Organism
Qualitative QC
NNRL, Y-1400 [CBS 142, CCRC 21672, CCY 17-14-1, IFO 0378, IFO 0610, JCM 2334, MUCL 30400, NCYC 445, VKM Y-2223]
Naganishia albida var. albidus
Fungi
5 Loops/Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Culti-Loops™ Naganishia albida var. albidus ATCC™ 10666™ > Naganishia albida var. albidus ATCC 10666
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.