Learn More
Thermo Scientific™ Culti-Loops™ Meyerozyma guilliermondii ATCC™ 6260™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4601521
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Candida species | |
Meyerozyma guilliermondii | |
Culti-Loops Quality Control Organism | |
Qualitative QC | |
ATCC 7375, CBS 566, DBVPG 6140, IGC 2730, JCM 1539, NRRL Y-324] |
6260TM* | |
Culti-Loops | |
Fungi | |
5 Loops/Pack |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.