missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ CTC LEAP PAL COMBI-xt HDHT Headspace Syringes

Operate leak-free thanks to a dynamic seal created by a spring-in-plunger design. Hamilton™ CTC LEAP PAL COMBI HDHT Headspace Syringes feature the high dynamic HD plunger which compensates for different expansion coefficients, creating a better seal over a larger range of temperatures.

Brand:  Hamilton™ 209685

 View more versions of this product

Product Code. 15356209

  • £260.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Performs over a large temperature range up to 200°C
  • Needle attachment is chemically inert, eliminating detached needles due to contact with organic, and chlorinated solvents
  • Needle attachment minimizes ghost peaks for wider range of sample components to be analyzed
  • Spring-in-plunger design creates dynamic seal between plunger tip and inside of the glass barrel for leak-free operation
  • Available in volumes ranging from 1 to 5mL
Specifications

Specifications

Not Autoclavable
0.0133 in.
0.025 in.
2 in.
Gastight, Headspace Syringe
392°F
68°F
20°C
5 mL
CTC LEAP CombiPAL Autosampler
23 G
0.34 mm
0.64 mm
50.8 mm
29 psig (2 bar)
200°C
1005 GF
1/Pk.
5
Glue-Free
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.