missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cresyl Violet perchlorate, 98%, pure
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Melting Point | 331.0°C to 333.0°C |
CAS | 41830-80-2 |
Assay Percent Range | 98% |
Molecular Formula | C16H12ClN3O5 |
Fluorescence Max | 615nm (in methanol) |
MDL Number | MFCD00012671 |
Synonym | Oxazine 9 perchlorate |
Solubility Information | Solubility in water: insoluble. |
InChI Key | HLHUTAPXZITNPJ-UHFFFAOYSA-N |
Infrared Spectrum | Authentic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'