missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresyl Violet acetate, pure, certified, Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 140.0°C to 143.0°C |
Absorption Ratio | 0.94 to 1.2 |
Dye Content | 65% min. |
Solubility Information | Solubility in water: soluble in water. Other solubilities: soluble in ethanol, insoluble in aceton |
Infrared Spectrum | Authentic |
IUPAC Name | acetic acid;5-iminobenzo[a]phenoxazin-9-amine |
Lambda Max | 596 to 599nm |
Formula Weight | 321.34 |
Grade | Pure |
Packaging | Glass bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'