missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red sodium salt, 0.04% w/v aq. soln.

Chemical Identifiers

CAS 62625-29-0
Molecular Formula C21H17O5S
MDL Number MFCD00001618
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11395547
SDS View Documents
Thermo Scientific Chemicals
042752.AE
100 mL N/A
N/A
11376149
SDS View Documents
Thermo Scientific Chemicals
042752.AP
500 mL N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62625-29-0
C21H17O5S
MFCD00001618

Specifications

62625-29-0
C21H17O5S
404.42
Cresol Red sodium salt
0.04% w/v aq. soln.
MFCD00001618
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 263-654-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.