missing translation for 'onlineSavingsMsg'
Learn More

winward engineering Calorimeter copper 100mm x 90mm 100mm x 90mm

Brand:  winward engineering XHM-310-050W

Calorimeter copper 100mm x 90mm 100mm x 90mm

Code : 02

Additional Details : Weight : 1.00000kg

Product Code. 13126310

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers