Learn More
Greiner Bio-One Conical Bottom Polypropylene Test Tubes with Screw Cap
For centrifugation and applications that require chemical and thermo resistant vessels.
£161.00 - £260.00
Specifications
Color Natural
Material Polypropylene
Bottom Shape Conical
View More Specs
Products 2
Product Code Brand Capacity (Metric) Closure Color Sterility Product Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Color Sterility Product Type Price Quantity & Availability  
10751624
View Documents
Greiner Bio-One
227285
50 mL White Sterile Centrifuge Tube
£260.00
Pack of 500
10529121
View Documents
Greiner Bio-One
210270
50 mL Blue Sterile Centrifuge Tube with Support Skirt
£161.00
Pack of 300
Description
Description
  • Natural colored tube
  • Blue graduations
  • White writing area
  • With or without support skirt
Specifications
Specifications
Natural
Polypropylene
Conical