missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25237-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene binds coiled-coil domain-containing protein 22 (CCDC22), and this complex can regulate the turnover of I-kappa-B and the activation of NF-kappa-B.Specifications
COMMD8 | |
Polyclonal | |
Unconjugated | |
COMMD8 | |
COMM domain containing 8, COMMD8, MDS022 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54951 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NX08 | |
COMMD8 | |
COMMD8 Fusion Protein Ag18712 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |