Learn More
Tenak™ Color Inserts for SlimTube™ Cryogenic Vials
Provide quick and easy sample identification. Tenak™ Color Inserts for SlimTube™ Cryogenic Vials fit snugly in the top of the SlimTube cap.
Brand: Tenak™ TE78415
Additional Details : Weight : 0.40000kg
Description
- Available in MonoColor (one color) or DualColor (two different colored sides)
- Packaged in bags of 100
- For use with SlimTube Cryogenic Vials
Specifications
MonoColor Green | |
500 | |
Round |
SlimTube cryogenic vials | |
5 bags of 100 |