missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hydroxide, 95%, pure, Thermo Scientific Chemicals

Product Code. 15209846
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 21041-93-0
Molecular Formula CoH2O2
Molecular Weight (g/mol) 92.95
MDL Number MFCD00016015
InChI Key ASKVAEGIVYSGNY-UHFFFAOYSA-L
Synonym cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
PubChem CID 10129900
SMILES [OH-].[OH-].[Co++]

Especificaciones

Quantity 1 kg
Assay Percent Range 95%
Linear Formula Co(OH)2
Merck Index 15,2426
IUPAC Name cobalt(2+);dihydroxide
Formula Weight 92.95
Percent Purity 95%
Grade Pure
Chemical Name or Material Cobalt(II) hydroxide
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Cobalt(II) hydroxide
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'