missing translation for 'onlineSavingsMsg'
Learn More

Alpha Packaging™ Clear Polyethylene Waste Sacks

Brand:  Alpha Packaging™ 13132390

Code : 03

Additional Details : Weight : 18.00000kg

Product Code. 13132390

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Waste Sack
100Pack
60 μm
1371 mm
Clear
Polyethylene
610 x 1054mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers