Learn More
Hirschmann™ Class B Graduated Polypropylene Cylinders with Raised Blue Graduations
Excellent for precise liquid measurement and fluid transfer in the laboratory.
Brand: Hirschmann™ 7500194
This item is not returnable.
View return policy
Description
- Acc. to DIN 12681/ISO 6706
- PP
- Raised scale
- Main point ring graduation
- Hexagonal base
Specifications
B | |
Round | |
Blue | |
482 mm |
Reusable | |
Precise Liquid Measurement and Fluid Transfer in the Laboratory | |
Hexagonal | |
Graduated Measuring Cylinder |