missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Class 3.3 Independent Temperature Safety Device

For Use With: KBF 115

Brand:  BINDER™ 8012-1032

Product Code. 16464029

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Class 3.3
  • With visual alarm
  • Safety devices also available in Class 2 and 3.1
Specifications

Specifications

Class 3.3 independent temperature safety device - with visual alarm (DIN 12880)
Temperature Safety Device
Visual alarm
BINDER Chambers KBF 115
Temperature Regulation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers