missing translation for 'onlineSavingsMsg'
Learn More

CLASP1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA564205

Product Code. 15751972

  • £447.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: TSPTNCSHGG LSPSRLWGWS ADGLAKHPPP FSQPNSIPTA PSHKALRRSY SPSMLDYDT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLASP1
Polyclonal
Unconjugated
CLASP1
1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1
Rabbit
Antigen affinity chromatography
RUO
23332
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7Z460
CLASP1
Recombinant protein corresponding to Human CLASP1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only