missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Mouse anti-Human, Mouse, Pig, Rat, Clone: 2A7D11, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen Chromogranin A
Clone 2A7D11
Concentration 1000 μg/mL
Applications Immunohistochemistry, Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17761129
View Documents
Proteintech
60135-2-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
17751129
View Documents
Proteintech
60135-2-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human, Mouse, Pig, Rat samples. It is validated for IHC, WB,ELISA
Specifications

Specifications

Chromogranin A
1000 μg/mL
Monoclonal
Liquid
Human, Mouse, Pig, Rat
1113
Primary
Store at -20°C
2A7D11
Immunohistochemistry, Western Blot, ELISA
Unconjugated
Mouse
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
IgG1
Protein G purification
Antibody
Videos
SDS
Documents

Documents

Product Certifications