missing translation for 'onlineSavingsMsg'
Learn More

chroman-8-carbaldehyde, 95%, Thermo Scientific™

£65.30 - £168.00

Chemical Identifiers

CAS 327183-32-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD12198125
InChI Key ZYKTXRDSAFQAAM-UHFFFAOYSA-N
Synonym chroman-8-carbaldehyde, 3,4-dihydro-2h-1-benzopyran-8-carbaldehyde, chromane-8-carbaldehyde, 2h-1-benzopyran-8-carboxaldehyde, 3,4-dihydro
PubChem CID 22265336
IUPAC Name 3,4-dihydro-2H-chromene-8-carbaldehyde
SMILES C1CC2=CC=CC(=C2OC1)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10184104
SDS View Documents
Thermo Scientific Maybridge
CC76104CB
250 mg Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10144154
SDS View Documents
Thermo Scientific Maybridge
CC76104DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

327183-32-4
162.188
ZYKTXRDSAFQAAM-UHFFFAOYSA-N
22265336
C1CC2=CC=CC(=C2OC1)C=O
C10H10O2
MFCD12198125
chroman-8-carbaldehyde, 3,4-dihydro-2h-1-benzopyran-8-carbaldehyde, chromane-8-carbaldehyde, 2h-1-benzopyran-8-carboxaldehyde, 3,4-dihydro
3,4-dihydro-2H-chromene-8-carbaldehyde

Specifications

327183-32-4
100.0
C10H10O2
chroman-8-carbaldehyde, 3,4-dihydro-2h-1-benzopyran-8-carbaldehyde, chromane-8-carbaldehyde, 2h-1-benzopyran-8-carboxaldehyde, 3,4-dihydro
C1CC2=CC=CC(=C2OC1)C=O
162.188
162.19
Chroman-8-carbaldehyde
94.5
95%
MFCD12198125
ZYKTXRDSAFQAAM-UHFFFAOYSA-N
3,4-dihydro-2H-chromene-8-carbaldehyde
22265336
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.