Learn More
Thermo Scientific™ Chromacol Vial Racks
Ensure a safe working position on the lab bench and during transport with these robust vial racks.
Brand:  Thermo Scientific™ T-25
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11922585
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Save space in the fridge while ensuring a safe working position on the bench and during transport. Available in a various materials, colors and capacities, Thermo Scientific™ Chromacol Vial Racks meet a variety of application needs
Choose from an assortment of chemically resistant and fairly robust racks.
  • Aluminum and plastic models are temperature resistant from -90 to +121ºC (-130 to +250ºF)
  • Aluminum and plastic models are autoclavable
Specifications
Specifications
Vial Rack
Autoclavable
Vials
25
Vial Rack
Blue
Aluminum
12 mm OD Vials
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.