missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriethylsilane, 99%, Thermo Scientific Chemicals

£49.85 - £394.00

Chemical Identifiers

CAS 994-30-9
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00000507
InChI Key DCFKHNIGBAHNSS-UHFFFAOYSA-N
Synonym triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
PubChem CID 13819
IUPAC Name chloro(triethyl)silane
SMILES CC[Si](Cl)(CC)CC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10216800
View Documents
Thermo Scientific Acros
215940250
25 mL Glass bottle
£49.85
25mL
Please to purchase this item. Need a web account? Register with us today!
10032870
View Documents Promotion Details
Thermo Scientific Acros
215941000
100 mL Glass bottle
£117.00
100mL
Please to purchase this item. Need a web account? Register with us today!
11955821
View Documents
Thermo Scientific Acros
215945000
500 mL Glass bottle
£394.00
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
Specifications

Chemical Identifiers

994-30-9
150.72
DCFKHNIGBAHNSS-UHFFFAOYSA-N
13819
CC[Si](Cl)(CC)CC
C6H15ClSi
MFCD00000507
triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
chloro(triethyl)silane

Specifications

994-30-9
0.8900g/mL
29°C
98.5% min. (GC)
(C2H5)3SiCl
MFCD00000507
13,165
0.89
Solubility in water: decomposes
CC[Si](Cl)(CC)CC
150.72
150.72
Liquid
Colorless to Yellow
142.0°C to 144.0°C
Authentic
C6H15ClSi
1.4300 to 1.4320
04,627
15,2179
triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
DCFKHNIGBAHNSS-UHFFFAOYSA-N
chloro(triethyl)silane
13819
99%
Chlorotriethylsilane
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 213-615-6

Product Identifier
  • Chlorotriethylsilane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only