missing translation for 'onlineSavingsMsg'
Learn More

Chlorogenic acid, Thermo Scientific Chemicals

Product Code. 15433229
Click to view available options
Quantity:
250 mg
1 g
5 g

An analog of caffeic acid

Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.

Solubility
Soluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-97-9
Molecular Formula C16H18O9
Molecular Weight (g/mol) 354.31
MDL Number MFCD00003862
InChI Key CWVRJTMFETXNAD-JUHZACGLSA-N
Synonym chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri
PubChem CID 1794427
ChEBI CHEBI:16112
IUPAC Name (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O

Specifications

Melting Point 210°C (decomposition)
Quantity 5 g
Merck Index 14,2142
Solubility Information Soluble in ethanol and acetone.
Formula Weight 354.31
Physical Form Powder
Chemical Name or Material Chlorogenic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'