missing translation for 'onlineSavingsMsg'
Learn More

Chlorocarbonylsulfenyl chloride, 97%, Thermo Scientific Chemicals

£17.29 - £41.90

Chemical Identifiers

CAS 2757-23-5
Molecular Formula CCl2OS
Molecular Weight (g/mol) 130.97
MDL Number MFCD00000703
InChI Key MNOALXGAYUJNKX-UHFFFAOYSA-N
Synonym chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
PubChem CID 75990
IUPAC Name S-chloro chloromethanethioate
SMILES ClSC(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362516
View Documents
Thermo Scientific Alfa Aesar
L06432.03
1 g
£17.29
1g
Please to purchase this item. Need a web account? Register with us today!
11372516
View Documents
Thermo Scientific Alfa Aesar
L06432.06
5 g
£41.90
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2757-23-5
130.97
MNOALXGAYUJNKX-UHFFFAOYSA-N
75990
ClSC(Cl)=O
CCl2OS
MFCD00000703
chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
S-chloro chloromethanethioate

Specifications

2757-23-5
CCl2OS
UN3265
chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
ClSC(Cl)=O
130.97
130.98
Chlorocarbonylsulfenyl chloride
98°C to 101°C
MFCD00000703
506318
MNOALXGAYUJNKX-UHFFFAOYSA-N
S-chloro chloromethanethioate
75990
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only