Learn More
WTW™ Chemical Oxygen Demand (COD) Test Kits
Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.
Brand: WTW™ 250309
Additional Details : CAS Number : 7783-35-9 Weight : 0.59400kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
- Safe determinations of COD in water
- Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
- For waste water, production control, ground water, surface water
- For PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, spectroFlex photometers
Specifications
For waste water, production control, ground water, surface water | |
Certificates | |
6 at 20°C |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers | |
200 | |
Chlorine dioxide (ClO2) reagent test kit |
Safety and Handling
- COD Cell Test Method: photometric 500 - 10000 mg/l Spectroquant
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Germ cell mutagenicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Reproductive toxicity Category 1B
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H311 + H331-Toxic in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, potassium dichromate