missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15528224
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Density Gradient Centrifugation; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Quantity 1 kg
Formula Weight 168.36
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 3.99kg
Chemical Name or Material Cesium Chloride
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cesium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'