missing translation for 'onlineSavingsMsg'
Learn More

Cesium carbonate, 99.9% (metals basis), Thermo Scientific Chemicals

£28.44 - £412.05

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331458
SDS View Documents
Thermo Scientific Alfa Aesar
010924.06
5 g
£28.44
5g
Please to purchase this item. Need a web account? Register with us today!
11311458
SDS View Documents
Thermo Scientific Alfa Aesar
010924.14
25 g
£45.73
25g
Please to purchase this item. Need a web account? Register with us today!
11321458
SDS View Documents
Thermo Scientific Alfa Aesar
010924.22
100 g
£125.63
100g
Please to purchase this item. Need a web account? Register with us today!
11341458
SDS View Documents
Thermo Scientific Alfa Aesar
010924.36
500 g
£412.05
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

534-17-8
325.82
FJDQFPXHSGXQBY-UHFFFAOYSA-L
10796
CCs2O3
MFCD00010957
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
[Cs+].[Cs+].[O-]C([O-])=O

Specifications

610°C (decomposition)
Powder/Granules
CCs2O3
4546405
Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide.
[Cs+].[Cs+].[O-]C([O-])=O
325.82
325.82
4.072
534-17-8
99.9% (metals basis)
MFCD00010957
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
FJDQFPXHSGXQBY-UHFFFAOYSA-L
dicaesium(1+) carbonate
10796
Hygroscopic
Cesium carbonate
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c

H318-H361f-H373

missing translation for 'einecsNumber' : 208-591-9

missing translation for 'rtecsNumber' : FK9400000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.