missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate

Color: Clear; Sterility: Sterile; Lid: Without Lid

Brand:  Greiner Bio-One 655162

 View more versions of this product

Product Code. 13002203

  • £286.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Compatible with automated systems
  • Alphanumeric well coding
  • High clarity of microplates for optimal microscopic examinations
  • Stackable
  • Individu
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

Free of detectable DNase/RNase, human DNA and pyrogens; contents non-cytotoxic
Without Lid
8 x 12
5/Pk., 100/Cs.
Polystyrene
50 to 335 μL
Cell Culture
4800 x g (swinging bucket rotor)
5
-20°C to +60°C
128 x 85 x 15 mm
Standard
34 mm2
Not Autoclavable
Clear
96
Sterile
Flat Bottom (F-well)
100
100/Cs.
Chimney
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers