Learn More
Greiner Bio-One CELLCOAT™ Poly-L-Lysine Protein Coated Flask
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ CELLCOAT™ Poly-L-Lysine Protein Coated Flask helps in promotion of cell adhesion and proliferation.
Brand:  Greiner Bio-One 658930
Product Code. 11823171
  • £1155.00 / Pack of 50
Explore more special offers
Description
Description
Poly-L-Lysine (PLL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
  • Coated with Poly-L-Lysine synthetic protein (30-70 kDa)
  • For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
  • Ready-to-use products
  • Includes lid with condensation rings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Canted
Non-sterile
250 mL
Poly-L-Lysine
Filter Screw Cap