missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD98/SLC3A2 Mouse anti-Human, Unconjugated, Clone: 2B10F5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CD98/SLC3A2 |
---|---|
Clone | 2B10F5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829155
|
Proteintech
66883-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819155
|
Proteintech
66883-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD98/SLC3A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD98/SLC3A2. This antibody recognizes human antigen. The CD98/SLC3A2 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
CD98/SLC3A2 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
6520 | |
Primary |
2B10F5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Human | |
IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title