Learn More
CD56 Mouse anti-Human, PE, Clone: MEM 188, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech PE-65067-100TESTS
This item is not returnable.
View return policy
Description
The CD56 antibody from Proteintech is a mouse monoclonal antibody generated with acute myelogenous leukemia cell line kg-1 of human CD56. This antibody recognizes human antigen. The CD56 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
CD56 | |
Monoclonal | |
10 μL/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
MEM 188 | |
PE | |
100 Tests | |
4684 | |
IgG2a |