Learn More
CD106 Mouse anti-Human, FITC, Clone: 1.G11B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech FITC-65049-100TESTS
Description
Vascular cell adhesion molecule 1 (VCAM1), also known as CD106, is a 110-kDa transmembrane glycoprotein belonging to the immunoglobulin gene superfamily. VCAM1 is expressed by cytokine-activated endothelium, interacts with integrin VLA4 (α4β1) present on the surface of leukocytes, and mediates both adhesion and signal transduction. It is also expressed either constitutively or inducibly in a variety of other cell types, including vascular smooth muscle cells, differentiating skeletal muscle cells, renal and neural epithelial cells, macrophages (Kupffer cells), dendritic cells, and bone marrow stromal cells (PMID: 7507076, 11359843).
Specifications
CD106 | |
Monoclonal | |
10 μL/Test | |
Mouse | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
1.G11B1 | |
FITC | |
100 Tests | |
7412 | |