missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cathepsin K Substrate (fluorogenic)

For cellular and molecular biology applications

£109.55 - £209.04

Specifications

Color White
Form Solid
For Use With (Application) A sensitive fluorogenic substrate for cathepsin K
Substrate Type Cathepsin Substrate
Molecular Weight (g/mol) 578.65
View More Specs

Products 2
Product Code Brand Substrate Quantity Price Quantity & Availability  
Product Code Brand Substrate Quantity Price Quantity & Availability  
15446189
View Documents
Thermo Scientific Chemicals
J64082.MA
10mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15456189
View Documents
Thermo Scientific Chemicals
J64082.MB
25mg
£209.04
25mg
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
Description

Description

A sensitive fluorogenic substrate for cathepsin K

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White
A sensitive fluorogenic substrate for cathepsin K
578.65
Solid
Cathepsin Substrate
-30° to -10°C
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.