missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carmine, High Purity Biological Stain (Alum Lake of Carminic Acid)
£24.92 - £259.29
Chemical Identifiers
CAS | 1390-65-4 |
---|---|
Molecular Formula | C22H20O13 |
Molecular Weight (g/mol) | 492.39 |
MDL Number | MFCD00167028 |
InChI Key | DGQLVPJVXFOQEV-UHFFFAOYNA-N |
Synonym | C.I. 75470 |
PubChem CID | 25113282 |
IUPAC Name | 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid |
SMILES | CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10194060
|
Thermo Scientific Chemicals
190200050 |
5 g | Glass bottle |
£24.92
5g |
In Stock
Log in to see stock. |
||||
10666962
|
Thermo Scientific Chemicals
190200250 |
25 g | Glass bottle |
£73.16
25g |
In Stock
Log in to see stock. |
||||
10360494
|
Thermo Scientific Chemicals
190201000 |
100 g | Glass bottle |
£259.29
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1390-65-4 | |
492.39 | |
DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
25113282 | |
CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O |
C22H20O13 | |
MFCD00167028 | |
C.I. 75470 | |
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid |
Specifications
1390-65-4 | |
MFCD00167028 | |
C.I. 75470 | |
DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O | |
492.39 | |
20% max. | |
Pure | |
Purple/Red to Red | |
Carmine |
C22H20O13 | |
15, 2442 | |
Solubility in water: soluble in hot water. Other solubilities: soluble in alkali, insoluble in diluted acids | |
Authentic | |
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid | |
25113282 | |
≥42.0% (carminic acid) | |
Complies | |
Powder |
Safety and Handling
Product Identifier
- Carmine
RUO – Research Use Only