Learn More
Zeiss™ Digital Camera Adapter P95 M37/52x0.75 for Primo Star™ and Primo Vert™ Microscopes
For connecting commercial digital still and video cameras to microscopes
Brand:  Zeiss™ 415500-1812-000
Product Code. 10464085
  • £296.00 / Each
Estimated Shipment: 15-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Zeiss Microscopes
Specifications
Specifications
Digital Camera Adapter
Accepts cameras with filter threads of M37mm or M52mm with a pitch of 0.75mm
Primo Star™ and Primo Vert Microscopes,
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Zeiss™ Digital Camera Adapter P95 M37/52x0.75 for Primo Star™ and Primo Vert™ Microscopes > Digital Camera Adapter