missing translation for 'onlineSavingsMsg'
Learn More

(Carbethoxymethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

£44.72 - £419.09

Chemical Identifiers

CAS 1099-45-2
Molecular Formula C22H21O2P
Molecular Weight (g/mol) 348.38
MDL Number MFCD00009183
InChI Key IIHPVYJPDKJYOU-UHFFFAOYSA-N
Synonym carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
PubChem CID 70670
IUPAC Name ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10496900
View Documents Promotion Details
Thermo Scientific Acros
108290250
25 g Glass bottle
£44.72
25g
Please to purchase this item. Need a web account? Register with us today!
10020600
View Documents
Thermo Scientific Acros
108291000
100 g Glass bottle
£121.61
100g
Please to purchase this item. Need a web account? Register with us today!
11955791
View Documents
Thermo Scientific Acros
108295000
500 g Glass bottle
£419.09
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

1099-45-2
348.38
IIHPVYJPDKJYOU-UHFFFAOYSA-N
70670
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C22H21O2P
MFCD00009183
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate

Specifications

1099-45-2
100.0
White to Yellow
98% min. (HPLC)
(C6H5)3P=CHCO2C2H5
01,112; 02,60; 10,78
Solubility in water: insoluble
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
348.38
348.38
Powder
98.0
124.0°C to 129.0°C
Authentic
C22H21O2P
MFCD00009183
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
IIHPVYJPDKJYOU-UHFFFAOYSA-N
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
70670
98+%
(Carbethoxymethylene)triphenylphosphorane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 214-151-7

Product Identifier
  • (Carbethoxymethylene)triphenylphosphorane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.