missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals CAPSO, 99%
£26.23 - £553.76
Chemical Identifiers
CAS | 73463-39-5 |
---|---|
Molecular Formula | C9H19NO4S |
Molecular Weight (g/mol) | 237.3 |
InChI Key | INEWUCPYEUEQTN-UHFFFAOYSA-N |
Synonym | capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid |
PubChem CID | 2733480 |
IUPAC Name | 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid |
SMILES | C1CCC(CC1)NCC(CS(=O)(=O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11975421
|
Thermo Scientific Chemicals
327740250 |
25 g | Glass bottle |
£26.23
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11985421
|
Thermo Scientific Chemicals
327741000 |
100 g | Glass bottle |
£71.46
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965421
|
Thermo Scientific Chemicals
327740010 |
1 kg | Plastic bottle |
£553.76
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
73463-39-5 | |
237.3 | |
capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid | |
3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid |
C9H19NO4S | |
INEWUCPYEUEQTN-UHFFFAOYSA-N | |
2733480 | |
C1CCC(CC1)NCC(CS(=O)(=O)O)O |
Specifications
73463-39-5 | |
263.0°C to 271.0°C | |
5 to 6.5 (1% aq. soln. at 25°C) | |
99% | |
capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid | |
INEWUCPYEUEQTN-UHFFFAOYSA-N | |
3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid | |
2733480 | |
99% | |
CAPSO |
(0.1M aq. soln.), 0.03 max. at 280nm, (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm | |
White | |
Authentic | |
C9H19NO4S | |
(0.1M in water) Clear colorless solution | |
C1CCC(CC1)NCC(CS(=O)(=O)O)O | |
237.3 | |
237.3 | |
Powder |
Safety and Handling
Product Identifier
- CAPSO
RUO – Research Use Only