Learn More
Thermo Scientific™ Capitol Vial Specimen Collection Beakers
Capitol Vial Specimen Beakers for easy sample collection and transfer.
£185.00 - £325.00
Product Code | Brand | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
15628558
|
Thermo Scientific™
10121 |
3.5 oz. | 103.5 mL | Pour Spout |
£218.00
Pack of 350 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15638558
|
Thermo Scientific™
10126 |
8 oz. | 236 mL | Temperature Strip |
£185.00
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15648558
|
Thermo Scientific™
10141 |
3.5 oz. | 103.5 mL | Temperature Strip |
£325.00
Pack of 350 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ Capitol Vial Specimen Collection Beakers are offered in two volumes, 3 oz. and 8 oz., to support a variety of sample collection needs. The beakers feature pour spouts for easy sample transfer.
- Molded pour spouts enable easy sample transfer
- Beakers are available with or without temperature indicator strips
- Collection cup holders are compatible with 8 oz. beakers
- Beakers and holders are constructed from virgin polypropylene